Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
$68.53 - $68.53
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-925-4
thermo scientific™
21230010
4L 100mm Each for $68.53
Case of 6 Each for $411.18
Description & Specifications
Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.
  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.