Thermo Scientific™ Nalgene™ Lab Notebooks with PolyPaper™ Pages
Permanently secure valuable notes and data in these hardcover case-bound paginated books featuring 100 (50 sheets) uncoated Thermo Scientific™ PolyPaper™.
$213.27 - $213.27
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
11-900 Thermo Scientific™
N63001000
0.25 in. grid; Black polyethylene hardcover Each for $213.27
Case of 6 Each for $1,279.62
Specifications
Description
Permanently secure your valuable notes and data in Thermo Scientific™ Nalgene™ Lab Notebooks with PolyPaper™ Pages. These hardcover case-bound paginated books include 100 (50 sheets) of uncoated PolyPaper and are extremely chemical resistant. Pages include spaces for dates and signatures — necessary documentation for securing patent protection.
  • Case-bound polyethylene hardcover protects 100 permanently-bound, polyolefin pages that are chemical resistant.
  • 9.25 x 11.25″ (23 x 29cm) pages are ruled with a fine green 0.25″ (6mm) grid to assist for orderly note keeping, drawings and charts.
  • Pages accepts ballpoint pen and permanent marking pen.
  • Notebook is permanently bound and paginated for data security and traceability.
Includes:
Instructions for keeping records, page for issuing book and table of contents
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.