Thermo Scientific™ Nalgene™ Lab Carrier
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers.
$337.03 - $337.03
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-251-1 Thermo Scientific™
N71310001
Lab Carrier Each for $337.03
Case of 2 Each for $674.06
Specifications
Description
Reduce breakage and spillage when transporting glass and plastic bottles by using Thermo Scientific™ Nalgene™ Lab Carriers. The carrier used without inserts accommodates square and rectangular bottles or Nalgene™ Unwire™ test tube racks.
  • Remove sleeves to accommodate up to six bottles with 2.375″ (6cm), 3.5″ (9cm), or 4.25″ (10.8cm) diameters
  • Sturdy, high density polyethylene construction holds six full 1000mL glass media bottles
  • Carriers stack when empty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.