Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ L900 Liquid Detergent
Thoroughly clean all plastic labware, with an environmentally friendly formula that will not cause crazing, stress cracking or clouding when used as directed.
$92.29 - $92.29
Products
Catalog Number Mfr. No. Description Price Quantity    
04-319B
thermo scientific™
N9004000
L900 Liquid Detergent Each for $92.29
Case of 4 Each for $369.16
Description & Specifications
Thoroughly clean all plastic labware, especially polycarbonate, with Thermo Scientific™ Nalgene™ L900 Liquid Detergent. This environmentally friendly formula will not cause crazing, stress cracking or clouding of plastic when used as directed.
  • For hand or machine use
  • Biodegradable
  • Non-toxic
  • Low foaming
  • Specially formulated for Nalgene plastic labware
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.