Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Heavy-Duty Wide-Mouth HDPE Jug with Closure
Store solids or powders in the Thermo Scientific™ Nalgene™ Heavy-Duty Wide-Mouth HDPE Jug with Closure, featuring the largest opening in the Nalgene carboy line.
$351.43 - $351.43
Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    
02-960-11
thermo scientific™
22410050
20L 120mm Each for $351.43
Case of 4 Each for $1,405.72
Description & Specifications
Store solids or powders in the Thermo Scientific™ Nalgene™ Heavy-Duty Wide-Mouth HDPE Jug with Closure, featuring the largest opening in the Nalgene carboy line. The wide, 4″ high-density Polyethelyne mouth allows for simplified cleaning and easy filling/emptying. This stable, space-efficient jug also features a molded-in handle and recessed bottom for easy handling and reduced spills.
Features:
  • Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing.
  • Wide stance and low center of gravity adds stability.
  • Integral shoulder handle and molded bottom grip make lifting comfortable.
  • Space-saving rectangular shape permits efficient storage.
  • Includes white, polypropylene screw closure.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.