Thermo Scientific™ Nalgene™ Heavy-Duty Rectangular HDPE Carboy
Store and transport reagents with this rugged, rectangular-shaped carboy.
$1,687.13 - $1,687.13
Disclaimers:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity  
02-961-56A Thermo Scientific™
N22140050
70mm 20L Case of 4 Each for $1,687.13
Specifications
Description
Store and transport reagents with the rugged Thermo Scientific™ Nalgene™ Heavy-Duty HDPE Carboy. The rectangular shape helps optimize limited storage space, and the large handle is easy to use, even with gloved hands. This heavy-duty polyethelyne carboy is both durable and space-saving.
Features:
  • Heavy walled construction for durability and chemical resistance
  • Graduated to 5L and 1 gal. increments
  • Valve-sealing graduated/leakproof† linerless 70mm screw closure
  • Not Autoclavable
Note: These tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.