Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ HDPE Vacuum Check Valve and Positive Connector
Prevent back pressure with aspirator-type vacuum pumps using the Thermo Scientific™ Nalgene™ Vacuum Check Valve with Pressure Connector.
$167.32 - $167.32
Products
Catalog Number Mfr. No. Description Price Quantity    
15-339-2
thermo scientific™
N61200010
Vacuum Check Valve and positive connector Pack of 6 Each for $167.32
Case of 12 Pack for $2,007.84
Description & Specifications
Prevent back pressure with aspirator-type vacuum pumps using the Thermo Scientific™ Nalgene™ Vacuum Check Valve and Pressure Connector. Designed for use with 1/4″ (6.35mm) to 5/16″ (7.94mm) ID tubing, they work best between 10- and 28- in. Hg.
Features:
  • Two-piece design is easy to disassemble for cleaning
  • Silicone diaphragm prevents back pressure
Note:
Not for use with liquids.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.