Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ HDPE Jerricans with Closure
Pour easily and accurately with the angled integral spout on Thermo Scientific™ Nalgene™ HDPE Jerricans.
$136.73 - $222.40
Specifications
Closure Color White
Closure Material Polypropylene 
Closure Type Screw
Material HDPE
Neck Finish 53Bmm
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    
02-961-50A
thermo scientific™
N22400015
6L 53Bmm Each for $136.73
Case of 6 Each for $820.38
02-961-50B
thermo scientific™
N22400025
10L 53Bmm Each for $152.53
Case of 6 Each for $915.18
02-961-50C
thermo scientific™
N22400050
20L 53Bmm Each for $222.40
Case of 4 Each for $889.60
Description & Specifications
Specifications
Closure Color White
Closure Material Polypropylene 
Closure Type Screw
Material HDPE
Neck Finish 53Bmm
No. per Pack 1
Pour easily and accurately with the angled integral spout of Thermo Scientific™ Nalgene™ HDPE Jerricans, featuring rugged design with wide stance and low center of gravity for greater stability. A molded-in handle and recessed bottom provide a sturdy grip when lifting and pouring while the space-saving rectangular shape optimizes limited storage space.
Key Features:
  • Integral spout long enough for accurate pouring
  • Attached, white polypropylene 53B closure won't get lost
  • Molded in graduations in 1 gal. and 5L increments for ease in dispensing
  • Graduated/Leakproof†
Includes:
White polypropylene screw closure
Warranty: 90 days