Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack


Store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure.

Manufacturer: thermo scientific™  3620089025

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 02-923-142

  • / Case of 2000 Each
 

Description & Specifications

Specifications

Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition.
Capacity (English) 0.25 oz.
Diameter (Metric) 25mm
Diameter (Metric) Inner Neck 14mm
Height (Metric) 45mm
Neck Finish 20-415mm
No. per Case 2000
Capacity 0.25 oz., 8mL
Capacity (Metric) 8mL
No. per Pack 2000
Diameter (Metric) 25mm
Height (Metric) 45mm

Store samples and ship small volumes of valuable reagents and light sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof†, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).

Features:

  • Nonsterile
  • Durable, uniform walls are resistant to splitting or puncturing
  • Leakproof† when assembled with Nalgene closure (must be ordered separately)

Ordering Information:

Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).