Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure: Nonsterile, Tray Pack


Perform many small volume applications with these safe and leakproof diagnostic bottles, ideal for use in test kits, sample storage and reagent shipping.

Manufacturer: thermo scientific™  N3220029025

 View more versions of this product

 Disclaimers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 02-930-153

  • / Case of 1500 Each
 

Description & Specifications

Specifications

Capacity 0.25 oz., 8mL
Capacity (Metric) 8mL
Capacity (English) 0.25 oz.
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Diameter (Metric) Inner Neck 14mm
No. per Case 1500
No. per Pack 150
Diameter (Metric) 25mm
Height (Metric) 45mm
Neck Finish 20-415mm

Perform many small volume applications with safe and leakproof† Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles with Closure. These bottles are ideal for use in test kits, sample storage and shipping of small volume, valuable reagents. They come packed in rigid trays for easy handling.

  • Nonsterile
  • Natural translucent HDPE with Natural PP Closure provides excellent chemical resistance
  • Durable, uniform walls are resistant to splitting or puncturing
  • Bottles are packed in rigid trays allowing for easy handling
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays. Closures are packed in a separate polybag within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.