Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Graduated HDPE Bucket
Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle.
$256.90 - $256.90
Products
Catalog Number Mfr. No. Description Price Quantity    
03-687-30
thermo scientific™
N70020025
2.5 gal. (9.5L) Bucket Each for $256.90
Case of 6 Each for $1,541.40
Description & Specifications
Transfer laboratory solutions and powders with the Thermo Scientific™ Nalgene™ Graduated HDPE Bucket, featuring a wide-lipped pour spout and polyethylene-coated handle. This heavy-duty, rugged high density polyethylene bucket is chemically compatible with a wide range of lab reagents.
  • High-density polyethylene construction offers more durability than hard rubber
  • Polyethylene-coated wire bail handle provides a comfortable and sturdy grip
  • Molded-in graduations in pints and liters are easy to read
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.