Thermo Scientific™ Nalgene™ Glass Prefilters
Increase filtration speed and throughput of highly viscous or heavily particulated solutions when using theses prefilters with Nalgene MF75 filter units or bottle top filters.
$286.38 - $390.03
Specifications
Material Glass-fiber
No. per Case 100
No. per Pack 100
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outer Price Quantity  
09-740-23 Thermo Scientific™
DS02815000
50mm Case of 100 Each for $286.38
09-740-24 Thermo Scientific™
DS02817500
75mm Case of 100 Each for $390.03
Specifications
Description
Increase filtration speed and throughput of highly viscous or heavily particulated solutions by using Thermo Scientific™ Nalgene™ Glass Fiber Prefilter Discs in conjunction with any Nalgene™ MF75 filter units or bottle top filters.
  • Glass fiber prefilter discs
  • For Nalgene 50, 75 and 90mm dia. reusable filtration products Cat. Nos. 300, DS0310, DS0330, DS0315, DS0320
Ordering Information:
The 75mm size is included in cases of Nalgene 500 and 1000mL nylon membrane and surfactant-free cellulose acetate (SFCA) membrane filter units.
Warranty: 90 days
Description & Specifications
Specifications
Glass-fiber
100
100