Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Fluorinated HDPE Solvent Wash Bottle
Eliminate risk of leakage with self-venting Thermo Scientific™ Nalgene™ Fluorinated HDPE Solvent Wash Bottles, which keep the solvent in the bottle.
$55.53 - $55.53
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
03-409-17B
thermo scientific™
N24210500
53mm 500mL Pack of 2 Each for $55.53
Case of 12 Pack for $666.36
Description & Specifications
Eliminate risk of leakage with self-venting Thermo Scientific™ Nalgene™ Fluorinated HDPE Solvent Wash Bottles, which keep the solvent in the bottle. Offering low permeability and excellent chemical resistance to many organic solvents, these high density polyethylene bottles are useful over a temperature range from 0° to 90°C. Bottles are designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The tip is molded to provide a straight stream with precision control and can be cut back to increase flow. Tight fit of the draw tube and closure prevents separation when squeezing bottle.
  • Polypropylene draw tube
  • Translucent fluorinated HDPE wash bottle with red polypropylene closure and stem
  • Bright red lettering silk-screened on bottle reads “Solvent Wash Bottle” for easy identification and no mix-ups
  • Low permeability and excellent chemical resistance to many organic solvents used in HPLC, TLC, and gas chromatography
  • Useful over a temperature range from 0° to 90°C (32° to 194°F)
  • Wide mouth designed for easy filling
  • Leakproof†
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.