Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Fluorinated HDPE Carboys with Closure
Enhance long-term container performance, prevent permeation loss and yield lower extractables. A cost-effective alternative to costly fluoropolymer containers.
$236.84 - $336.48
Specifications
Closure Size 83B
Material HDPE
Wide Mouth No
No. per Pack 1
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Capacity (Metric) Price Quantity    
03-312-18
thermo scientific™
20970020
10L Each for $236.84
Case of 6 Each for $1,421.04
03-312-19
thermo scientific™
20970050
20L Each for $336.48
Case of 4 Each for $1,345.92
Description & Specifications
Specifications
Closure Size 83B
Material HDPE
Wide Mouth No
No. per Pack 1
Enhance long-term container performance, prevent permeation loss and yield lower extractables with Thermo Scientific™ Nalgene™ Fluorinated Carboys with Closure, for use with most acids, alkalis and aggressive organic solvents. These safe, durable and cost-efficient carboys featuring fluorinated surfaces (inside and outside), for improved barrier properties and reduced solvent absorption and penetration. Fluorination enhances long-term container performance and prevents or reduces permeation loss.
  • Leakproof†
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • HDPE screw caps included
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days