Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Conical Oak Ridge Graduated Polypropylene Centrifuge Tube
Carry out applications with this tube, which combines the superior separation and pelleting of a conical tube with the sealing capability of an Oak Ridge tube.
$232.03 - $232.03
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
05-579-33
thermo scientific™
31480050
35mL 24mm Pack of 10 Each for $232.03
Case of 10 Pack for $2,320.30
Description & Specifications
Carry out a variety of applications with these tubes, which combine the superior separation and pelleting of a conical tube with the higher sealing capability of an Oak Ridge tube. Thermo Scientific™ Nalgene™ Conical Oak Ridge Graduated Polypropylene Centrifuge Tubes can spin up to 50,000 x g in a refrigerated or non-refrigerated centrifuge, conical tube adapter required.
Features:
  • Feature silk-screened graduations from 5 to 35mL in 1mL increments
  • Can withstand forces to 50,000 x g in refrigerated or non-refrigerated centrifuges—requires adapter, DS3147-0050
  • Supplied with polypropylene linerless closures for leak-proof performance up to 10,000 x g
  • Offer superior clarity
  • Polypropylene copolymer tubes are translucent and offer better chemical resistance than polycarbonate; repeatedly autoclavable
  • Tubes must be filled to at least 80% capacity for proper performance.
  • Autoclavable/Biohazard/Graduated/Leakproof†
Ordering Information:
To assure leakproof service, especially at speeds greater than 10,000 x g or when spinning hazardous materials, use Nalgene sealing cap Cat. No. DS3132-0024 or sealing cap assembly Cat. No. DS3131-0024.
Includes:
Supplied with linerless, polypropylene screw closure which will not leak during ordinary use.
Requires:
Both tubes require white polycarbonate tube adapter (see DS3147 0050 below) to centrifuge.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.