Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings
Use Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings to customize bottle and carboy closures for aseptic transfer of liquids.
$142.13 - $142.13
Specifications
Description Closure with Barbed Bulkhead Fitting
Neck Finish 38-430mm
For Use With Nalgene culture vessels
Material Polypropylene (PP)
No. per Case 4
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Inner Tubing Price Quantity    
02-923-14W
thermo scientific™
NDS21670001
1/2 in. Case of 4 Each for $142.13
02-923-14X
thermo scientific™
NDS21670002
1/4 in. Case of 4 Each for $142.13
Description & Specifications
Specifications
Description Closure with Barbed Bulkhead Fitting
Neck Finish 38-430mm
For Use With Nalgene culture vessels
Material Polypropylene (PP)
No. per Case 4
No. per Pack 4
Use Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings to customize most Nalgene bottle and carboy closures for aseptic transfer of liquids.
  • Size 38mm-430 closures are predrilled allowing for easy, safe in-house assembly
  • Barbed at both ends to accept tubing inside and outside the container
  • Molded of white, autoclavable polypropylene, ready for in-house sterilization
Includes: 
Instructions, drilling template and a set of two panel-mounted barbed fittings, acetal nuts and silicone gaskets.
Ordering Information: 
Barbed bulkhead fittings also sold separately (6149-0001 and 6149-0002).