Please login to your online account to display your discounted pricing

Thermo Scientific Nalgene™ Carboy Replacement Screw Closures and Gaskets


Guarantee a leakproof system using Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets.

Manufacturer: thermo scientific  N7121510053

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 12-565-978

  • / Case of 12 Each
 

Description & Specifications

Specifications

No. per Case 12
No. per Pack 12
Closure Size 53B
Color White
Description Replacement Closure
For Use With Nalgene Large Carboys and Bottles
Material HDPE

Guarantee leakproof services with secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys.

  • Made to fit Nalgene Carboys