Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure: Bulk Pack
Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure
$595.03 - $2,798.63
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Type Yes (bulk packed, unassembled)
Closure Color Natural
Closure Liner Linerless
Closure Material Polypropylene (PP)
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
02-923-37
thermo scientific™
3120030001
20mm 30mL Case of 1000 Each for $2,798.63
03-313-429
thermo scientific™
N3120030002
20mm 60mL Case of 1000 Each for $2,695.97
03-313-38C
thermo scientific™
N3120030004
24mm 125mL Case of 500 Each for $1,771.47
36-101-6764
thermo scientific™
N3120030016
28mm 500mL Case of 125 Each for $926.03
03-313-430
thermo scientific™
N3120030032
38-430mm 1L Case of 100 Each for $595.03
Description & Specifications
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Type Yes (bulk packed, unassembled)
Closure Color Natural
Closure Liner Linerless
Closure Material Polypropylene (PP)
Description Narrow-Mouth Boston Round Bottle
Color Natural
Material LDPE
Mouth Narrow
Shape Round
Package, ship and store a variety of reagents using Thermo Scientific™ Nalgene™ Boston Round Narrow-Mouth LDPE Bottles with Closure, constructed from natural low-density polyethylene.
Key Features
  • Durable and clear
  • Uniform walls that resist splitting and puncturing
  • Leakproof† when used with their Nalgene closures
  • Ideal for trace metal analysis—LDPE is low in trace metal content
Warranty: 90 days