Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Boston Round HDPE Bottles without Closure: Bulk Pack
Conduct demanding packaging applications with Thermo Scientific™ Nalgene™ Boston Round HDPE Bottles without Closure, which are both durable and reliable products.
$620.21 - $2,266.53
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Type Ordered separately
Description Boston Round Bottle
Color Natural
Material HDPE
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures
Products
Catalog Number Mfr. No. Closure Size Capacity (Metric) Price Quantity    
03-313-424
thermo scientific™
N3620020001
20mm 30mL Case of 1000 Each for $2,206.50
03-313-425
thermo scientific™
N3620020002
20mm 60mL Case of 1000 Each for $2,266.53
03-313-426
thermo scientific™
N3620020008
24mm 250mL Case of 250 Each for $1,148.97
03-313-427
thermo scientific™
N3620020016
28mm 500mL Case of 125 Each for $828.85
03-313-428
thermo scientific™
N3620020032
38-430mm 1L Case of 50 Each for $620.21
Description & Specifications
Specifications
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure Type Ordered separately
Description Boston Round Bottle
Color Natural
Material HDPE
Mouth Narrow
Shape Round
Conduct demanding packaging applications with Thermo Scientific™ Nalgene™ Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Features:
  • Natural, HDPE
  • Good for freezer use to -100°C
  • Leakproof†, when assembled with Nalgene closure (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. Order colored closures separately (Cat. No. 362150).
Warranty: 90 days