Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ BioTransport Carrier
Easily view contents with the Thermo Scientific™ Nalgene™ BioTransport Carrier.
$273.52 - $273.52
Products
Catalog Number Mfr. No. Description Price Quantity    
15-251-2
thermo scientific™
N71350001
BioTransport Carrier Each for $273.52
Case of 4 Each for $1,094.08
Description & Specifications
Transport racks containing tubes or sample holders filled with potentially infectious materials using the Thermo Scientific™ Nalgene™ BioTransport Carrier, which offers excellent visibility of contents.
  • Polycarbonate carrier is tough and break resistant
  • Easy to grasp, molded-in handles
  • Silicone gasket seals tightly
  • Clamps securely hold carrier closed and assure leak-resistant seal
  • Stainless-steel handle available separately (7136-0001)
  • For accidental spills inside of carrier, autoclave with clamps closed
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.