Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings
Customize tubing sets as culture vessels for aseptic transfer of liquids with Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings.
$44.23 - $52.13
Specifications
No. per Case 24
No. per Pack 2
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Tubing Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Tubing Price Quantity  
15-315-1 Thermo Scientific™
N61490001
0.5 in. Pack of 2 Each for $52.13
Case of 12 Pack for $625.56
15-315-2 Thermo Scientific™
N61490002
0.25 in. Pack of 2 Each for $44.23
Case of 12 Pack for $530.76
Specifications
Description
Customize tubing sets as culture vessels for aseptic transfer of liquids with autoclavable Thermo Scientific™ Nalgene™ Barbed Polypropylene Bulkhead Fittings, suitable for use with most Nalgene bottle and carboy closures. Pre-drilled closures allow easy, safe in-house assembly.
Features:
  • Closures accept (two) 0.25″ fittings
  • Fittings, barbed at both ends, accept tubing for inside and outside container
Includes:
Instructions, drilling template and two each of polypropylene barbed fitting, acetyl nuts, silicone gaskets and and TPE port caps.
Warranty: 90 days
Description & Specifications
Specifications
24
2