missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation

Ensure more complete drainage with the low outlet of these polypropylene carboys, excellent for distilled water and culture media.

$1,490.61 - $1,806.00

Specifications

Description Autoclavable Carboy w/Bottom Tubulation
Diameter (Metric) Inner Neck 64mm
Autoclavable Yes
Closure Size 83B
Graduations Molded-in graduations in 1 gal. and 5L increments
View More Specs

Alert:

Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity  
02-960-22 Thermo Scientific™
23010020
10L 83B Case of 6 Each for $1,806.00
02-690-23 Thermo Scientific™
23010050
20L 83B Case of 4 Each for $1,490.61
Specifications

Description

Ensure more complete drainage with the low outlet of Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation, excellent for distilled water and culture media.

Features:
  • Bottom tubulation molded in one piece with bottle
  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5L increments
  • Neck accepts No. 13.5 rubber stoppert
  • Autoclavable/Graduated/Leakproof†

Ordering Information:

Both sizes accept 0.5in. I.D. (1.3cm) tubing - Not Included

Includes:

White polypropylene screw cap with TPE gasket

Note:

A hose clamp should be used when attaching tubing to ensure tight fit. Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.

Warranty: 90 days.  For warranty terms, please contact technicalsupport@thermofisher.com

Description & Specifications

Specifications

Autoclavable Carboy w/Bottom Tubulation
Yes
Molded-in graduations in 1 gal. and 5L increments
Polypropylene
64mm
83B
Molded-in
1