Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with 1.5 in. Sanitary Flanges
Use these carboys as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.
$1,255.10 - $1,527.03
Specifications
Certifications/Compliance USP Class VI, non-cytotoxic
Description Autoclavable Carboy Sanitary Flange
Autoclavable Yes
Closure Size 83B
Graduations Graduated to contain in liters and gallons
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-963-3A
thermo scientific™
N26400020
10L 83B Each for $1,255.10
02-963-3B
thermo scientific™
N26400050
20L 83B Each for $1,273.39
02-963-3C
thermo scientific™
N26400130
50L 83B Each for $1,527.03
Description & Specifications
Specifications
Certifications/Compliance USP Class VI, non-cytotoxic
Description Autoclavable Carboy Sanitary Flange
Autoclavable Yes
Closure Size 83B
Graduations Graduated to contain in liters and gallons
Handles Molded-in
Material Polypropylene
Spigot 1-1/2 in. secure, non-threaded sanitary dispensing port
Use the Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with 1.5″ Sanitary Flanges as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.
Key Features:
  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5L
  • USPVI/Autoclavable/Graduated/Leakproof†
Includes:
Size 83B closure; accepts No. 13.5 rubber stopper.
Note: 
Follow autoclave instructions provided with the product for proper function.
Warranty: 90 days