Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack
Securely hold three Imhoff cones with this durable and transparent settling cone rack, featuring a wide base for stability on the lab bench.
$551.90 - $551.90
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
15-443-60 Thermo Scientific™
N10010010
Cone Rack Each for $551.90
Case of 4 Each for $2,207.60
Specifications
Description
Securely hold three Imhoff cones with the durable and transparent Thermo Scientific™ Nalgene™ Acrylic Imhoff Cone Rack, featuring a wide base for stability on the lab bench.
Features:
  • Transparent; easily see level of contents
  • Can also be used to hold 500 and 1000mL Nalgene separatory funnels (Cat. Nos. 4300 and 4301) for glass separatory funnels
Settling Cone Graduation Intervals
  • 0-1mL: 0.1mL
  • 1-10mL: 0.5mL
  • 10-20mL: 1mL
  • 20-40mL: 2mL
  • 40-100mL: 10mL
  • 100-500mL: 50mL
  • 500-1000mL: 100mL
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.