Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier
Secure bottles and reduce chance of bumping, cracking or chipping with this safety bottle carrier.
$323.77 - $323.77
Products
Catalog Number Mfr. No. Description Price Quantity    
03-439-31
thermo scientific™
65050010
Carrier Safety Bottle LDPE 500mL Each for $323.77
Case of 6 Each for $1,942.62
Description & Specifications
Secure bottles and reduce chance of bumping, cracking, or chipping. Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier offers greater rigidity and impact resistance. The sturdy, wide handle is molded in one piece with the base allowing a comfortable grip, even when the carrier is full. This linear low density polyethylene carrier holds up to six standard 500mL reagent bottles in any profile.
  • Cover latches securely to base and allow carriers to stack
  • Dimensions: 14L × 10W × 11″H (35 × 25 × 28cm)
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.