Thermo Scientific™ Nalgene™ 4mm Syringe Filters
Filter sample volume sizes of 0.5 to 1.0mL using Nalgene 4mm Syringe Filters.
$594.83 - $594.83
Specifications
Diameter 4mm
No. per Case 400
No. per Pack 100
View More Specs
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Membrane Pore Size Price Quantity  
Catalog Number Mfr. No. Membrane Pore Size Price Quantity  
09-740-34A Thermo Scientific™
N1710020
Cellulose Acetate 0.2μm Pack of 100 Each for $594.83
Case of 4 Pack for $2,379.32
09-740-34B Thermo Scientific™
N1710045
Cellulose Acetate 0.45μm Pack of 100 Each for $594.83
Case of 4 Pack for $2,379.32
09-740-34C Thermo Scientific™
N1760020
Nylon 0.2μm Pack of 100 Each for $594.83
Case of 4 Pack for $2,379.32
09-740-34D Thermo Scientific™
N1760045
Nylon 0.45μm Pack of 100 Each for $594.83
Case of 4 Pack for $2,379.32
Specifications
Description
Filter sample volume sizes of 0.5 to 1.0mL using Thermo Scientific™ Nalgene™ 4mm Syringe Filters. CA membrane filters are excellent for buffers and aqueous solutions, and EIA and ELISA assay sample prep. Nylon membrane filters are excellent for filtration of aqueous solutions, alcohols, DMSO, and HPLC and CG samples and solvents.
Cellulose Acetate (CA)
  • Low protein binding
  • Ideal for filtration of biological/immunological samples, cell culture media components, biological fluids, most other aqueous solutions and reagents, including EIA and ELISA samples
Nylon
  • Naturally hydrophilic, low extractables
  • Useful for filtering cryoprotectants such as DMSO and with aqueous/biological and nonaqueous/HPLC/GC samples and solvents
Warranty: 90 days unless otherwise stated.
Description & Specifications
Specifications
4mm
400
100