Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ 230mm Ceramic Metal Desiccator Plate
Ensure greater resistance to thermal shock than porcelain using this guaranteed unbreakable and corrosion-resistant ceramic metal desiccator plate.
$149.94 - $149.94
Products
Catalog Number Mfr. No. Description Price Quantity    
08-642-10
thermo scientific™
53120230
Each for $149.94
Case of 6 Each for $899.64
Description & Specifications
Ensure greater resistance to thermal shock than porcelain using the guaranteed unbreakable and corrosion-resistant Thermo Scientific™ Nalgene™ 230mm Ceramic Metal Desiccator Plate.
  • 230mm diameter fits the Nalgene Classic Dessicator (5311-0250) as well as the 250mm Nalgene Blue Desiccators (5309-0250 and 5310-0250)
  • Numbered quadrants for easy location of crucibles and other containers
  • Six holes in each quadrant and a 3mm center hole to enhance airflow and drying efficiency
  • Autoclavable
Ordering Information:
Individually packaged
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.