Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ 13mm Syringe Filters

Filter aqueous solutions and media or solvents in sample volumes from 2 to 10mL using Thermo Scientific™ Nalgene™ 13mm Syringe Filters.

$772.60 - $948.69

Specifications

Diameter (Metric) 13mm
Volume (Metric) 2 to 10mL
No. per Case 100
No. per Pack 100
View More Specs
Products
Catalog Number Mfr. No. Housing Material Membrane Pore Size Price Quantity    

09-740-100

 
thermo scientific™
N7201320
Polypropylene (PP) housing w/green ring PES 0.2μm Case of 100 Each for $948.69

09-740-101

 
thermo scientific™
N7211320
Polypropylene (PP) housing w/white ring PTFE 0.2μm Case of 100 Each for $772.60

09-740-102

 
thermo scientific™
N7211345
Polypropylene (PP) housing w/white ring PTFE 0.45μm Case of 100 Each for $772.60
Description & Specifications

Specifications

Diameter (Metric) 13mm
Volume (Metric) 2 to 10mL
No. per Case 100
No. per Pack 100

Filter aqueous solutions and media (PES membrane) or solvents (PTFE membrane) with Thermo Scientific™ Nalgene™ 13mm Syringe Filters. Accommodate sample volumes from 2 to 10mL.

  • 2 to 10mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip

PTFE

  • Naturally hydrophobic
  • Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housing
  • Excellent for HPLC/GC solvent filtration
  • Non-sterile; Autoclavable

Polyethersulfone (PES)

  • Higher flowrates, lower protein binding and lower extractables than cellulosic membranes
  • Ideal for sterile filtration and clarification of cell culture media, buffers and additives
  • Naturally hydrophilic; contains no wetting agents
  • Polypropylene housing
  • Certified sterile, non-cytotoxic and non-pyrogenic