Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ 13L HDPE Jerrican with Tethered Polypropylene Closure

Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure.

$699.03 - $858.41

Specifications

Capacity 3.5 gal., 13L
Capacity (English) 3.5 gal.
Certifications/Compliance USP Class VI Compliant/FDA21 CFR 177.1520 Compliant
Closure Color White
Closure Material Polypropylene (PP)
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Capacity (Metric) Neck Finish Price Quantity    

02-960-23

 
thermo scientific™
22430013
13L 53B Case of 4 Each for $699.03

02-960-24

 
thermo scientific™
22439013
13L 53B, 38-430mm Case of 4 Each for $858.41
Description & Specifications

Specifications

Capacity 3.5 gal., 13L
Capacity (English) 3.5 gal.
Certifications/Compliance USP Class VI Compliant/FDA21 CFR 177.1520 Compliant
Closure Color White
Closure Material Polypropylene (PP)
Closure Type Tethered
Capacity (Metric) 13L
Diameter (English) Inner Neck 1.5 in.
Diameter (Metric) Inner Neck 38mm
Material HDPE
Height (English) With Closure 15 in.
Height (Metric) With Closure 378mm
Length (English) 11 in.
Length (Metric) 290mm
No. per Case 4
No. per Pack 1
Width (Metric) 189mm

Store reagents with the Thermo Scientific™ Nalgene™ Jerrican with Tethered Polypropylene Closure, featuring a space-saving design, molded-in handle and recessed grip at the bottom to provide better control when pouring. The double-ported 2243-9013 is ideal as a reservoir in automated systems. The 53B closure can be plumbed with input/output tubing while the 38mm closure is used for refilling or emptying the reservoir.

Key Features:

  • Jerrican is ideal as a reservoir in automated systems like the PRISM™ 3700 DAN Analyzer
  • Molded-in graduations in 1 gal. and 5L increments allow for ease in dispensing
  • Molded-in handle provides a sturdy grip when carrying heavy contents
  • Integral shoulder handle and molded bottom grip make lifting comfortable
  • Wide stance and low center of gravity add stability
  • Heavy rugged design with good chemical resistance allows for a wide variety of solutions
  • Space-saving rectangular shape permits efficient storage

Ordering Information:

  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • Custom tubing sets can be designed at the Thermo Scientific Custom Tubing Set Configurator, www.thermoscientific.com/tubingconfigurator