Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Single-Use HDPE Carboy
Eliminate cleaning, prep and set-up costs with these ready-to-use sterile, one-time use HDPE carboys--ideal for storing sterile fluids and pharmaceutical/biotech products.
$499.87 - $1,168.84
Specifications
Certifications/Compliance Food-grade, USP Class VI, non-cytotoxic
Diameter (Metric) Inner Neck 64mm
Closure Size 83B
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
03-409-24B
thermo scientific™
N3422890050
20L 83B Case of 6 Each for $1,168.84
03-409-24C
thermo scientific
N3422890075
33L 83B Each for $499.87
Description & Specifications
Specifications
Certifications/Compliance Food-grade, USP Class VI, non-cytotoxic
Diameter (Metric) Inner Neck 64mm
Closure Size 83B
Eliminate cleaning, prep and set-up costs with ready-to-use Thermo Scientific™ Nalgene™ Single-Use HDPE Carboys. Supplied sterile for one-time use, the high-density polyethelyne vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.
Features
  • HDPE Carboy with polypropylene closure
  • Sterile
  • Leakproof†
Ordering Information
Attachable stainless-steel handle (Cat. No. 2229-0001) available for 20L single-use HDPE carboy (Cat. No. 342289-0050).
Includes
PP closure