Please login to your online account to display your discounted pricing

Naftopidil hydrochloride, Tocris

α1 antagonist

$73.75 - $299.87

Chemical Identifiers

Chemical Name or Material Naftopidil hydrochloride
Assay Percent Range >98%
Molecular Formula C24H28N2O3·HCl
Formula Weight 428.96
Synonym 4-(2-Methoxyphenyl)-α-[(1-naphthalenyloxy)methyl]-1-piperazineethanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

059710

 
r & d systems
059710
10mg Each for $73.75

059750

 
r & d systems
059750
50mg Each for $299.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 428.96
Synonym 4-(2-Methoxyphenyl)-α-[(1-naphthalenyloxy)methyl]-1-piperazineethanol hydrochloride
Chemical Name or Material Naftopidil hydrochloride
Assay Percent Range >98%
Molecular Formula C24H28N2O3·HCl
CAS 1164469-60-6
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature