Please login to your online account to display your discounted pricing

Naftopidil hydrochloride, Tocris


α1 antagonist

Quantity 50mg

Manufacturer: r & d systems  059750

 View more versions of this product

Catalog No. 059750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 428.96
Chemical Name or Material Naftopidil hydrochloride
Assay Percent Range >98%
Synonym 4-(2-Methoxyphenyl)-α-[(1-naphthalenyloxy)methyl]-1-piperazineethanol hydrochloride
Recommended Storage Store at Room Temperature
Molecular Formula C24H28N2O3·HCl
CAS 1164469-60-6

Specifications

Chemical Name or Material Naftopidil hydrochloride
Assay Percent Range >98%
Molecular Formula C24H28N2O3·HCl
Formula Weight 428.96
Synonym 4-(2-Methoxyphenyl)-α-[(1-naphthalenyloxy)methyl]-1-piperazineethanol hydrochloride
Accessories

Store at Room Temperature