Please login to your online account to display your discounted pricing

Alfa Aesar™ Nafion™ R-1100 resin, sulfonyl fluoride form

Thermoplastic-can be extruded or laminated

$56.64 - $2,722.99

Chemical Identifiers

Chemical Name or Material Nafion™ R-1100 resin
CAS 26654-97-7
Name Note Sulfonyl fluoride form
Molecular Formula Rf[OCF2CF(CF3)2]nOCF2CF2SO2F
MDL Number MFCD05665455
Synonym Perfluorosulfonyl fluoride-PTFE copolymer
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4462604

 
alfa aesar™
4462604
2g Each for $56.64

AA4462618

 
alfa aesar™
4462618
50g Each for $709.61

AA4462630

 
alfa aesar™
4462630
250g Each for $2,722.99
Description & Specifications

Specifications

Physical Form Pellet
Chemical Name or Material Nafion™ R-1100 resin
TSCA TSCA
Name Note Sulfonyl fluoride form
Molecular Formula Rf[OCF2CF(CF3)2]nOCF2CF2SO2F
MDL Number MFCD05665455
CAS 26654-97-7
Synonym Perfluorosulfonyl fluoride-PTFE copolymer
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >200°C
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.