missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nafion™ R-1100 resin, sulfonyl fluoride form

Thermoplastic-can be extruded or laminated

Manufacturer:  Alfa Aesar™ 4462609

 View more versions of this product

Catalog No. AA4462609



Specifications

Specifications

Nafion™ R-1100 resin
10g
26654-97-7
MFCD05665455
Insoluble in water.
C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)F)(F)F)F)(F)F)F.C(=C(F)F)(F)F
546.126
Pellet
>200°C
Sulfonyl fluoride form
C9F18O4S
1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-trifluoroethenyl oxy propan-2-yl oxy ethanesulfonyl fluoride-tetrafluoroethene 1:1
DHYYUWYUPUOPFL-UHFFFAOYSA-N
1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonyl fluoride
117323

Chemical Identifiers

26654-97-7
546.126
DHYYUWYUPUOPFL-UHFFFAOYSA-N
117323
C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)F)(F)F)F)(F)F)F.C(=C(F)F)(F)F
C9F18O4S
MFCD05665455
1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-trifluoroethenyl oxy propan-2-yl oxy ethanesulfonyl fluoride-tetrafluoroethene 1:1
1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonyl fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents