Please login to your online account to display your discounted pricing

Alfa Aesar™ Nafion™ NR50, super acidic ion exchange resin catalyst

Super acid catalyst

$165.74 - $165.74

Chemical Identifiers

Chemical Name or Material Nafion™ NR50
CAS 31175-20-9
Name Note Super acidic ion exchange resin catalyst
MDL Number MFCD00132790
Synonym Perfluorosulfonic acid-PTFE copolymer
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4080209

  SDS
alfa aesar™
4080209
10g Each for $165.74
Description & Specifications

Specifications

Physical Form Pellet
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to pale brown
Melting Point 200°C
TSCA TSCA
Name Note Super acidic ion exchange resin catalyst
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Density 2.1g/mL
Health Hazard 1 Warning
MDL Number MFCD00132790
Sensitivity Hygroscopic
CAS 31175-20-9
Quantity 10g
Chemical Name or Material Nafion™ NR50
Synonym Perfluorosulfonic acid-PTFE copolymer
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.