Please login to your online account to display your discounted pricing

Alfa Aesar™ Nafion™ D-521 dispersion, 5% w/w in water and 1-propanol, ≥0.92 meq/g exchange capacity

Preparation of thin films and coatings; proton exchange membranes

Chemical Identifiers

Chemical Name or Material Nafion™ D-521 dispersion
CAS 31175-20-9
Name Note ≥0.92meq/g exchange capacity
MDL Number MFCD00132790
UN Number UN1987
Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42117AC

  SDS
alfa aesar™
42117AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42117AE

  SDS
alfa aesar™
42117AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42117AP

  SDS
alfa aesar™
42117AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42117K4

  SDS
alfa aesar™
42117K4
2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25mL
Concentration 5% w/w in water and 1-propanol
Flash Point 24° (75°F)
CAS 31175-20-9
MDL Number MFCD00132790
Color Colorless to yellow
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 0.93g/mL
Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
UN Number UN1987
Physical Form Liquid
TSCA TSCA
Name Note ≥0.92meq/g exchange capacity
Chemical Name or Material Nafion™ D-521 dispersion
Health Hazard 2 GHS H Statement
H318-H226-H336
Causes serious eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H226-H336
Causes serious eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.