Please login to your online account to display your discounted pricing

Nafadotride, Tocris


Highly potent, preferential D3 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS134710

 View more versions of this product

Catalog No. 134710

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 365.47
Quantity 10mg
CAS 149649-22-9
Assay Percent Range >98%
Molecular Formula C22H27N3O2
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material Nafadotride
Synonym N-[(1-Butyl-2-pyrrolidinyl)methyl]-4-cyano-1-methoxy-2-naphthalenecarboxamide

Specifications

Chemical Name or Material Nafadotride
Assay Percent Range >98%
Molecular Formula C22H27N3O2
Formula Weight 365.47
Synonym N-[(1-Butyl-2-pyrrolidinyl)methyl]-4-cyano-1-methoxy-2-naphthalenecarboxamide
Accessories

Desiccate at Room Temperature