Please login to your online account to display your discounted pricing

Nafadotride, Tocris

Highly potent, preferential D3 antagonist

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material Nafadotride
Assay Percent Range >98%
Molecular Formula C22H27N3O2
Formula Weight 365.47
Synonym N-[(1-Butyl-2-pyrrolidinyl)methyl]-4-cyano-1-methoxy-2-naphthalenecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

134710

 
r & d systems
RDS134710
10mg Each for $257.85

134750

 
r & d systems
RDS134750
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 365.47
Quantity 10mg
CAS 149649-22-9
Chemical Name or Material Nafadotride
Assay Percent Range >98%
Molecular Formula C22H27N3O2
Synonym N-[(1-Butyl-2-pyrrolidinyl)methyl]-4-cyano-1-methoxy-2-naphthalenecarboxamide
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature