missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
NADPH tetrasodium salt hydrate, 96%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 328742500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
NADPH tetrasodium salt hydrate | |
2646-71-1 | |
100.0 | |
Glass bottle | |
MFCD10567216 | |
nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
WYWWVJHQDVCHKF-NRTBITFTNA-J | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium | |
131673986 | |
96% |
approx. 98% | |
97.5 | |
95% min. (HPLC) | |
C21H26N7Na4O17P3 | |
250 mg | |
Solubility in water: soluble | |
[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O | |
833.35 | |
833.35 | |
Extra Pure |
Chemical Identifiers
2646-71-1 | |
833.35 | |
WYWWVJHQDVCHKF-NRTBITFTNA-J | |
131673986 | |
[Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
C21H26N7Na4O17P3 | |
MFCD10567216 | |
nadph tetrasodium salt, nadph, tetrasodium salt, coenzyme ii tetrasodium salt reduced form | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium |
Safety and Handling
MOISTURE SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 220-163-3
RUO – Research Use Only