missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine Sulfate, TCI America™

Manufacturer:  TCI America E0657250G

 View more versions of this product

Catalog No. E0657250G



Specifications

Specifications

N4-Ethyl-N4-(2-hydroxyethyl)-2-methyl-1,4-phenylenediamine Sulfate
155°C
25646-77-9
2811
GVEYRUKUJCHJSR-UHFFFAOYSA-N
2-(4-amino-N-ethyl-3-methylanilino)ethanol;sulfuric acid
91579
Crystal-Powder at 20°C
White-Yellow
250g
C11H20N2O5S
kodak cd 4, 4-n-ethyl-n-2-hydroxyethyl-2-methylphenylenediamine sulfate, n4-ethyl-n4-2-hydroxyethyl-2-methyl-1,4-phenylenediamine sulfate, cd 4, ppd 4 4, 2-4-amino-3-methylphenyl ethylamino ethanol sulfate, 2-4-amino-3-methylphenyl ethyl amino ethanol sulfate, 2-4-amino-3-methylphenyl ethylamino ethanol sulfate, 4-ammonio-m-tolyl ethyl 2-hydroxyethyl ammonium sulphate, n5-ethyl-n5-2'-hydroxyethyl toluene-2,5-diammonium sulfate
CCN(CCO)C1=CC(=C(C=C1)N)C.OS(=O)(=O)O
292.35
292.35
≥98.0% (HPLC)

Chemical Identifiers

25646-77-9
292.35
kodak cd 4, 4-n-ethyl-n-2-hydroxyethyl-2-methylphenylenediamine sulfate, n4-ethyl-n4-2-hydroxyethyl-2-methyl-1,4-phenylenediamine sulfate, cd 4, ppd 4 4, 2-4-amino-3-methylphenyl ethylamino ethanol sulfate, 2-4-amino-3-methylphenyl ethyl amino ethanol sulfate, 2-4-amino-3-methylphenyl ethylamino ethanol sulfate, 4-ammonio-m-tolyl ethyl 2-hydroxyethyl ammonium sulphate, n5-ethyl-n5-2'-hydroxyethyl toluene-2,5-diammonium sulfate
2-(4-amino-N-ethyl-3-methylanilino)ethanol;sulfuric acid
C11H20N2O5S
GVEYRUKUJCHJSR-UHFFFAOYSA-N
91579
CCN(CCO)C1=CC(=C(C=C1)N)C.OS(=O)(=O)O
Safety and Handling

Safety and Handling

RTECSNumber : KJ6477000

TSCA : Yes

Documents