missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine, 98%

Chemical Identifiers

Molecular Formula C37H34FN3O7
MDL Number MFCD15145192
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64614MC View Documents Alfa Aesar™
J64614MC
100mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ6461403 View Documents Alfa Aesar™
J6461403
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

C37H34FN3O7
MFCD15145192

Specifications

White to Yellow
C37H34FN3O7
651.68
98%
100mg
MFCD15145192
Powder
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxycytidine
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C