missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-deoxycytidine 99.0+%, TCI America™
$262.13 - $769.78
Chemical Identifiers
CAS | 67219-55-0 |
---|---|
Molecular Formula | C37H35N3O7 |
Molecular Weight (g/mol) | 633.701 |
MDL Number | MFCD00010114 |
InChI Key | MYSNCIZBPUPZMQ-VOTWKOMSSA-N |
Synonym | Bz-DMT-dC |
PubChem CID | 2724490 |
IUPAC Name | N-[1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide |
SMILES | COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)OCC4C(CC(O4)N5C=CC(=NC5=O)NC(=O)C6=CC=CC=C6)O |
Chemical Identifiers
67219-55-0 | |
633.701 | |
MYSNCIZBPUPZMQ-VOTWKOMSSA-N | |
2724490 | |
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)OCC4C(CC(O4)N5C=CC(=NC5=O)NC(=O)C6=CC=CC=C6)O |
C37H35N3O7 | |
MFCD00010114 | |
Bz-DMT-dC | |
N-[1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide |
Specifications
67219-55-0 | |
White | |
MFCD00010114 | |
Bz-DMT-dC | |
COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=C(C=C3)OC)OCC4C(CC(O4)N5C=CC(=NC5=O)NC(=O)C6=CC=CC=C6)O | |
633.701 | |
633.70 | |
Crystalline Powder |
119°C | |
C37H35N3O7 | |
1 g | |
MYSNCIZBPUPZMQ-VOTWKOMSSA-N | |
N-[1-[(2R,4S,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-hydroxyoxolan-2-yl]-2-oxopyrimidin-4-yl]benzamide | |
2724490 | |
≥99.0% (HPLC) | |
N4-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxycytidine |
Safety and Handling
EINECSNumber : (5)-6799
TSCA : Yes