missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N2-Methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CD05460EA

 View more versions of this product

Catalog No. CD05460EA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

183.16
97%
Amber Glass Bottle
MFCD00042321
N2-Methyl-5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-amine
97%
10g
100.0
25366-22-7
96.5
C4H4F3N3S

Chemical Identifiers

25366-22-7
100.0
C4H4F3N3S
MFCD00042321
97%
96.5
97%
183.16
N2-Methyl-5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-amine

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.