Please login to your online account to display your discounted pricing

N1,N12-Diethylspermine tetrahydrochloride, Tocris

Polyamine synthase inhibitor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material N1, N12-Diethylspermine tetrahydrochloride
Assay Percent Range >98%
Molecular Formula C14H34N4·4HCl
Formula Weight 404.29
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

050010

 
r & d systems
RDS050010
10mg Each for $168.15

050050

 
r & d systems
RDS050050
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 404.29
Chemical Name or Material N1, N12-Diethylspermine tetrahydrochloride
Assay Percent Range >98%
Molecular Formula C14H34N4·4HCl
CAS 113812-15-0
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature