Please login to your online account to display your discounted pricing

N1,N11-Diethylnorspermine tetrahydrochloride, Tocris

Spermine and spermidine acetyltransferase potentiator

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material N1, N11-Diethylnorspermine tetrahydrochloride
Assay Percent Range >98%
Molecular Formula C13H32N4·4HCl
Formula Weight 390.27
Synonym N, N'-bis[3-(Ethylamino)propyl]-1, 3-propanediamine tetrahydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

046850

 
r & d systems
RDS046850
50mg Each for $746.48

046810

 
r & d systems
RDS046810
10mg Each for $177.37
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 390.27
CAS 156886-85-0
Assay Percent Range >98%
Synonym N, N'-bis[3-(Ethylamino)propyl]-1, 3-propanediamine tetrahydrochloride
Chemical Name or Material N1, N11-Diethylnorspermine tetrahydrochloride
Molecular Formula C13H32N4·4HCl
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature