Please login to your online account to display your discounted pricing

N1-Ethyl-4-(trifluoromethoxy)aniline, Technical Grade, Maybridge

Chemical Identifiers

Chemical Name or Material N1-Ethyl-4-(trifluoromethoxy)aniline
Assay Tech.
Molecular Formula C9H10F3NO
Linear Formula CF3OC6H4NHCH2CH3
Formula Weight 205.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00676EA

  SDS
fisher scientific company
TL00676EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.18
Purity Grade Technical Grade
Quantity 10g
Assay Tech.
Molecular Formula C9H10F3NO
Linear Formula CF3OC6H4NHCH2CH3
CAS 175278-20-3
Packaging Amber Glass Bottle
Chemical Name or Material N1-Ethyl-4-(trifluoromethoxy)aniline