Please login to your online account to display your discounted pricing

N1-(4-Butyryl-3-hydroxyphenyl)acetamide, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
Assay Tech.
Molecular Formula C12H15NO3
Formula Weight 221.25
MDL Number MFCD00276573
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB01739DA

  SDS
fisher scientific company
SB01739DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SB01739EA

  SDS
fisher scientific company
SB01739EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.25
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
Molecular Formula C12H15NO3
CAS 28583-62-2
MDL Number MFCD00276573
Packaging Amber glass bottle
Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide