Please login to your online account to display your discounted pricing

N1-(4-Butyryl-3-hydroxyphenyl)acetamide, Tech., Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01739EA

 View more versions of this product

Catalog No. SB01739EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 221.25
Purity Grade Technical Grade
Quantity 10g
Assay Tech.
CAS 28583-62-2
Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
MDL Number MFCD00276573
Packaging Amber glass bottle
Molecular Formula C12H15NO3

Specifications

Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
Assay Tech.
Molecular Formula C12H15NO3
Formula Weight 221.25
MDL Number MFCD00276573