Please login to your online account to display your discounted pricing

N1-(4-Butyryl-3-hydroxyphenyl)acetamide, Tech., Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB01739DA

 View more versions of this product

Catalog No. SB01739DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Amber glass bottle
Quantity 1g
Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
CAS 28583-62-2
Assay Tech.
Molecular Formula C12H15NO3
Formula Weight 221.25g/mol
MDL Number MFCD00276573

Specifications

Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
Assay Tech.
Molecular Formula C12H15NO3
Formula Weight 221.25g/mol
MDL Number MFCD00276573