Please login to your online account to display your discounted pricing

N1-(4-Butylphenyl)acetamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material N1-(4-Butylphenyl)acetamide
CAS 3663-20-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H17NO
Linear Formula CH3CH2CH2CH2C6H4NHC(O)CH3
Formula Weight 191.27
MDL Number MFCD00277211
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00828EA

  SDS
fisher scientific company
TL00828EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.27
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material N1-(4-Butylphenyl)acetamide
CAS 3663-20-5
MDL Number MFCD00277211
Packaging Amber Glass Bottle
Molecular Formula C12H17NO
Linear Formula CH3CH2CH2CH2C6H4NHC(O)CH3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.